Learn More
ERA WATERS CORP PAT700 CONDUCTIVITY SOLN KIT
SDP
Catalog No. NC1862937
Encompass
"(1) 100 uS/cm KCl in 60 mL vial with RFID Tag.This CRM is produced under ISO 9001, ISO/IEC 17025 and ISO 17034 (previously Guide 34) accredited quality system."
Catalog No. NC1862937 Supplier ERA WATERS CORP Supplier No. 02602ERA
"(1) 100 uS/cm KCl in 60 mL vial with RFID Tag.This CRM is produced under ISO 9001, ISO/IEC 17025 and ISO 17034 (previously Guide 34) accredited quality system."