Learn More
ERA WATERS CORP PAT700 CONDUCTIVITY STNDRD
SDP
Catalog No. NC0275486
Encompass
100 uS/cm KCl - 60 mL bottle with RFID Tag. Replaces FG7002602
Catalog No. NC0275486 Supplier ERA WATERS CORP Supplier No. 02602
100 uS/cm KCl - 60 mL bottle with RFID Tag. Replaces FG7002602