Learn More
ERA WATERS CORP PAT700 System Suitability Kit
SDP
Catalog No. NC1680971
Encompass
Set of three bottles includes (1) Reagent Water Rw, (1) 0.500 mg/L C USP Sucrose Rs, (1) 0.500 mg/L C 1,4-benzoquinone Rss in 60 mL vials with RFID Tags.
Catalog No. NC1680971 Supplier ERA WATERS CORP Supplier No. 18402
Add to Cart
Edge
Add to Cart
Set of three bottles includes (1) Reagent Water Rw, (1) 0.500 mg/L C USP Sucrose Rs, (1) 0.500 mg/L C 1,4-benzoquinone Rss in 60 mL vials with RFID Tags.