Learn More
ERA WATERS CORP PAT700 Validation Control Kit
SDP
Catalog No. NC1830732
Encompass
Set of two bottles includes (1) Blank, (1) 0.500 mg/L C NIST Sucrose in 60 mL bottles with RFID Tags.
Catalog No. NC1830732 Supplier ERA WATERS CORP Supplier No. 19222
Set of two bottles includes (1) Blank, (1) 0.500 mg/L C NIST Sucrose in 60 mL bottles with RFID Tags.