Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Potassium Hydroxide 0.05N in Methanol (1L)
SDP

Catalog No. NC9393809
Encompass

Potassium Hydroxide 0.05N in Methanol (1L)

Catalog No. NC9393809 Supplier Aqua Solutions Supplier No. TEN017
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Hydroxide 0.05N in Methanol (1L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.