Learn More
Warner Instruments™ Patch Clamp Glass, 8250
Catalog No. BTX640817 Shop All BTX Harvard Products
Encompass_Preferred
Change view
Click to view available options
Color:
Clear
Catalog No. Color
BTX640817 Clear
1 options
Catalog No. BTX640817 Supplier Warner Instruments™ Supplier No. 640817
8250 patch clamp glass measures 1.5 mm x 3 in. for patch clamp applications.
  • Same glass as Schott 8250
  • Seals were formed faster, and maintained for longer periods using 8250 glass
  • 8250 glass is manufactured without an inner filament
  • This glass is offered in both standard and premium models
  • The premium glass has fire-polished ends and is cleaned with deionized water before being packed in dust-free containers
  • Initial tests showed the Custom 8250 glass to be equal to the Corning 7052 (a favorite among researchers performing patch clamping) in noise performance.
Specifications
Material Borosilicate Glass
Sterility Non-sterile
Color Clear
Length (Metric) 75 mm
Disposable Limited-use