Learn More
Grainger Patch Cord Cat 6 Booted Green 14 ft.
Catalog No. 502751231
Encompass_Preferred
The Grainger Approved Cat 6 Booted Patch Cord is a high-quality network cable designed for fast Ethernet connections. It features Category 6 performance for speeds up to 10Gbps, four twisted pair 24 AWG stranded conductors, and snagless boots to protect the RJ45 connectors. This cord is UL verified, ETL listed, and complies with ANSI/TIA-568-C.2 standards. Offered in multiple lengths from 1 to 15 ft., this durable patch cable delivers reliable network connections.
Catalog No. 50-275-1231 Supplier Grainger Supplier No. 14A980
The Grainger Approved Cat 6 Booted Patch Cord is a high-quality network cable designed for fast Ethernet connections. It features Category 6 performance for speeds up to 10Gbps, four twisted pair 24 AWG stranded conductors, and snagless boots to protect the RJ45 connectors. This cord is UL verified, ETL listed, and complies with ANSI/TIA-568-C.2 standards. Offered in multiple lengths from 1 to 15 ft., this durable patch cable delivers reliable network connections.