Learn More
Grainger Patch Cord Cat 6 Booted Yellow 5.0 ft.
Catalog No. 502750083
Encompass_Preferred
The Monoprice Voice and Data Patch Cord is a Cat6 Ethernet cable that connects devices on a local area network. It has RJ45 connectors and is available in multiple lengths from 1 foot to 100 feet. The cord is UL rated, and constructed with 4 pair 24AWG stranded copper conductors, polyethylene insulation, and a PVC jacket. It supports 10BASE-T, 100BASE-TX, 1000BASE-T/1000BASE-TX and 10GBASE-T network speeds. This high quality, snagless patch cable provides reliable network connectivity.
Catalog No. 50-275-0083 Supplier Grainger Supplier No. 14A983
The Monoprice Voice and Data Patch Cord is a Cat6 Ethernet cable that connects devices on a local area network. It has RJ45 connectors and is available in multiple lengths from 1 foot to 100 feet. The cord is UL rated, and constructed with 4 pair 24AWG stranded copper conductors, polyethylene insulation, and a PVC jacket. It supports 10BASE-T, 100BASE-TX, 1000BASE-T/1000BASE-TX and 10GBASE-T network speeds. This high quality, snagless patch cable provides reliable network connectivity.