Learn More
Med Vet International Eye Patch Nexcare Opticlude, 1539, 20/Box
Catalog No. 502831447
Encompass_Preferred
Nexcare Opticlude eye patches are made of plastic and feature a breathable backing Gentle adhesive located around the border holds the eye patch in place yet allows for easy removal Hypoallergenic and latex-free for use on all skin types, including sensitive Contoured design provides ability to move eyelids for comfort Disposable eye patch available in regular size in a 20-count box
Catalog No. 50-283-1447 Supplier Med Vet International Supplier No. 5778BX
Nexcare Opticlude eye patches are made of plastic and feature a breathable backing Gentle adhesive located around the border holds the eye patch in place yet allows for easy removal Hypoallergenic and latex-free for use on all skin types, including sensitive Contoured design provides ability to move eyelids for comfort Disposable eye patch available in regular size in a 20-count box