Learn More
LEONI FIBER OPTICS INC PATCHCORD-8M-600/660/710 S/S
Catalog No. NC1523960
Encompass
Patchcord-8m-600/660/710 s/s 0.22NA LOH-SMA905 on both ends-SSTL BXStripwound-SMA/SMA adapter included
Catalog No. NC1523960 Supplier LEONI FIBER OPTICS INC Supplier No. 093389
Patchcord-8m-600/660/710 s/s 0.22NA LOH-SMA905 on both ends-SSTL BXStripwound-SMA/SMA adapter included