Learn More
Medchemexpress LLC Patchouli alcohol | 5986-55-0 | MFCD00209526 | 99.4% | 222.37 g·mol⁻¹ | C15H26O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN020710MG
Encompass_Preferred
Patchouli alcohol is a natural tricyclic sesquiterpene supplied for laboratory research and analytical use. It is reported to exhibit anti-Helicobacter pylori, anti-inflammatory, and antioxidant properties, and is provided at high purity for reproducible experimental results.
  • Natural tricyclic sesquiterpene with reported antibacterial, anti-inflammatory, and antioxidant activity.
  • High purity (~99.4%) suitable as an analytical or research standard.
  • Available in low-mass vials for analytical testing and method development.
  • Supplied as a solid with a defined molecular weight (222.37 g·mol⁻¹) and formula (C15H26O).
  • For research use only; not intended for human or animal therapeutic use.
Catalog No. 50-002-95494