Learn More
Medchemexpress LLC Patent Blue V 100mg
SDP
Catalog No. 5000206642
Encompass_Preferred
Patent Blue V (Acid blue 1) is a novel biological dye that can be used as an intraocular dye for retinectomy Retinectomy refers to the removal of the translucent inner limiting membrane (ILM) The application of appropriate dyes in vitreoretinal surgery can achieve the purpose of complete removal Patent Blue V can be used to stain retinal premembranous structures Spectral analysis shows that Patent Blue V has strong absorption below 450 nm and above 600 nm showing a blue-green color Patent Blue V is also used as a marker in lymphangiography for resection of neoplastic lymph nodes[1][2]
Catalog No. 50-002-06642 Supplier Medchemexpress LLC Supplier No. HYW110929100MG
Patent Blue V (Acid blue 1) is a novel biological dye that can be used as an intraocular dye for retinectomy Retinectomy refers to the removal of the translucent inner limiting membrane (ILM) The application of appropriate dyes in vitreoretinal surgery can achieve the purpose of complete removal Patent Blue V can be used to stain retinal premembranous structures Spectral analysis shows that Patent Blue V has strong absorption below 450 nm and above 600 nm showing a blue-green color Patent Blue V is also used as a marker in lymphangiography for resection of neoplastic lymph nodes[1][2]