Learn More
Medchemexpress LLC Patent blue V calcium salt | 3536-49-0 | 100.0% | 580.72 | 500 MG
SDP
Catalog No. 5000435779
Encompass_Preferred
Patent Blue V calcium salt is a triarylmethane dye for tissue staining and lymphatic tracing. Administered via topical injection or eye drops, it stains specific tissues like corneal endothelium and the lymphatic system to aid precise surgical operations. It finds application in ophthalmic surgery and sentinel lymph node biopsy for tumors. It's a potential alternative to Trypan blue for DMEK graft staining. Staining results are detectable by UV-visible spectrophotometry or fluorescence spectroscopy.
  • Tissue staining and lymphatic tracing.
  • Topical injection or eye drop administration.
  • Stains corneal endothelium and lymphatic system.
  • Aids precise surgical operations.
  • Used in ophthalmic surgery.
  • Used in sentinel lymph node biopsy.
  • Potential alternative to Trypan blue.
  • Detect by UV-Vis or fluorescence spectroscopy.
Catalog No. 50-004-35779 Supplier Medchemexpress LLC Supplier No. HY126395500MG
Patent Blue V calcium salt is a triarylmethane dye for tissue staining and lymphatic tracing. Administered via topical injection or eye drops, it stains specific tissues like corneal endothelium and the lymphatic system to aid precise surgical operations. It finds application in ophthalmic surgery and sentinel lymph node biopsy for tumors. It's a potential alternative to Trypan blue for DMEK graft staining. Staining results are detectable by UV-visible spectrophotometry or fluorescence spectroscopy.
  • Tissue staining and lymphatic tracing.
  • Topical injection or eye drop administration.
  • Stains corneal endothelium and lymphatic system.
  • Aids precise surgical operations.
  • Used in ophthalmic surgery.
  • Used in sentinel lymph node biopsy.
  • Potential alternative to Trypan blue.
  • Detect by UV-Vis or fluorescence spectroscopy.