Learn More
Electron Microscopy Sciences Paterson 35 mm Tank & Reel, Set
SDP
Catalog No. 5030496
Encompass_Preferred
The super System 4 is simpler to load, faster to fill, and easier to clean than any other tank. It has a large diameter one-piece lid and funnel, which clips into place positively and securely enabling the tank to be filled and emptied quickly. Supplied complete with one 35 mm/120/220 Auto Load Reel, Watertight EVA cap prevents leakage during inversion agitation. Uses less than 300 ml solution for a single 35 mm film. Takes a single 35mm film. Uses less than 300ml.
Catalog No. 50-304-96 Supplier Electron Microscopy Sciences Supplier No. 7434214
The super System 4 is simpler to load, faster to fill, and easier to clean than any other tank. It has a large diameter one-piece lid and funnel, which clips into place positively and securely enabling the tank to be filled and emptied quickly. Supplied complete with one 35 mm/120/220 Auto Load Reel, Watertight EVA cap prevents leakage during inversion agitation. Uses less than 300 ml solution for a single 35 mm film. Takes a single 35mm film. Uses less than 300ml.