Learn More
Electron Microscopy Sciences Paterson Auto Load Reel
SDP
Supplier:  Electron Microscopy Sciences 7433806
Encompass_Preferred
Designed for use in the Paterson Super System 4 tank, the Auto Load Reel allows thru free flow of solutions. Paterson Reels have a smooth finish and use the unique twin ball ratchet system. Film loading just couldn't be easier!
Catalog No. 50-304-95