Learn More
Electron Microscopy Sciences Paterson Force Film Washer, Set
SDP
Catalog No. 5030492
Encompass_Preferred
This quick and efficient film washing system fits most domestic taps. It is designed for the Paterson Super System 4 tanks.
Catalog No. 50-304-92 Supplier Electron Microscopy Sciences Supplier No. 7433601
This quick and efficient film washing system fits most domestic taps. It is designed for the Paterson Super System 4 tanks.