Learn More
Electron Microscopy Sciences Paterson Paterson Print Tong
SDP
Catalog No. 5031301
Encompass_Preferred
These tongs have textured side panels which offer a firm grip and deeply notched "shoulders" which avoid slippage into the tray. The tongs are all plastic and chemically resistant. Tongs are color-coded in light and dark gray to help minimize the risk of cross-contamination.
Catalog No. 50-313-01 Supplier Electron Microscopy Sciences Supplier No. 74817
These tongs have textured side panels which offer a firm grip and deeply notched "shoulders" which avoid slippage into the tray. The tongs are all plastic and chemically resistant. Tongs are color-coded in light and dark gray to help minimize the risk of cross-contamination.