Learn More
Electron Microscopy Sciences Paterson Print Washer
SDP
Catalog No. 5027727
Encompass_Preferred
The standard washer is perfect for washing both RC and fiber-based papers. To use, prints are loaded into the cradle which is then lowered into the tank. Faucet pressure gently rocks the print cradle as water continuously exchanges. All prints remain separate and water freely circulates. The washer holds up to 12-8x10" or 24-5x7" prints.
Catalog No. 50-277-27 Supplier Electron Microscopy Sciences Supplier No. 7435310
The standard washer is perfect for washing both RC and fiber-based papers. To use, prints are loaded into the cradle which is then lowered into the tank. Faucet pressure gently rocks the print cradle as water continuously exchanges. All prints remain separate and water freely circulates. The washer holds up to 12-8x10" or 24-5x7" prints.