Learn More
Electron Microscopy Sciences Paterson RC Rapid Drying Rack
SDP
Catalog No. 5030498
Encompass_Preferred
For air drying up to 20 RC prints in 15 min. Its compact design permits maximum ambient air circulation and prevents trapping of water between the print and the rack. The rack is also suitable for use with fiber-based paper. Print capacity: 5- 11x14"; 10-8x10"; 20-5x7".
Catalog No. 50-304-98 Supplier Electron Microscopy Sciences Supplier No. 74839
For air drying up to 20 RC prints in 15 min. Its compact design permits maximum ambient air circulation and prevents trapping of water between the print and the rack. The rack is also suitable for use with fiber-based paper. Print capacity: 5- 11x14"; 10-8x10"; 20-5x7".