Learn More
Electron Microscopy Sciences Paterson Rs Print Squeegee
SDP
Catalog No. 5027729
Encompass_Preferred
Extra soft rubber blades and a unique floating-action lower blade pre-vents any damage to the print surface. Suitable for both RC and fiber-based papers. The Print Squeegee handles prints up to 16x20" (40x50cm).
Catalog No. 50-277-29 Supplier Electron Microscopy Sciences Supplier No. 74823
Extra soft rubber blades and a unique floating-action lower blade pre-vents any damage to the print surface. Suitable for both RC and fiber-based papers. The Print Squeegee handles prints up to 16x20" (40x50cm).