Learn More
Electron Microscopy Sciences Paterson Darkroom Paterson Safelight, 'A' Dome
SDP
Catalog No. 5033317
Encompass_Preferred
Perfect for the small darkroom, or for safety spot lighting individual work areas. This compact safelight, 4"(D)x6 1/4"(H), can be installed wherever it's convenient - freestanding, mounted on the wall, or ceiling. The orange globe supplied is safe for all black-and-white papers.  The unit comes with a 15W lamp, 120V AC. 3-prong cord.
Catalog No. 50-333-17 Supplier Electron Microscopy Sciences Supplier No. 7432710
Perfect for the small darkroom, or for safety spot lighting individual work areas. This compact safelight, 4"(D)x6 1/4"(H), can be installed wherever it's convenient - freestanding, mounted on the wall, or ceiling. The orange globe supplied is safe for all black-and-white papers.  The unit comes with a 15W lamp, 120V AC. 3-prong cord.