Learn More
Dynamic Diagnostics Path Tray For Slides and Blocks
Catalog No. 22899668
Change view
Click to view available options
Dimensions (L x W x H):
13.25 x 8.875 x 0.75 in.
Catalog No. Dimensions (L x W x H)
22-899-668 13.25 x 8.875 x 0.75 in.
1 options
Catalog No. 22-899-668 Supplier Dynamic Diagnostics Supplier No. PT20
Add to Cart
Add to Cart
The PathTray hold slides, blocks and paperwork all in one tray while having the slides readily viewable. PathTrays are stackable and give a clean organized look. This tray is simple, smart and convenient.
  • Stackable and neatly organize paperwork for each case
  • Minimizes potential errors by keeping paperwork together
  • Slides are easily viewed by the pathologist
  • Improves organization and work flow
  • Minimizes space with a clean stackable appearance
  • Less chance for patient to patient mix up
  • Built for years of service
  • Xylene resistant and durable compared to cardboard
  • Easy to view slides
  • Paperwork is contained, stackable
  • Holds up to 20 slides and five tissue blocks
  • Xylene resistant
  • Lightweight and durable
Specifications
Dimensions (L x W x H) 13.25 x 8.875 x 0.75 in.
Material Polypropylene
For Use With Slides, paraffin blocks or cassettes