Learn More
Mopec PathCam Hands-Free Foot Pedal Command
SDP
Catalog No. 23987017 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Foot Pedal
Catalog No. For Use With (Equipment)
23-987-017 Foot Pedal
1 options
Catalog No. 23-987-017 Supplier Mopec Supplier No. AX047
Add to Cart
Add to Cart
Designed to allow the user to program four different pedals to the functions of their choosing.
  • Programmable quad USB foot switch
  • Side-by-side footswitch configuration
  • Simple plug-and-play USB compliant foot switch which uses standard USB HID drive
  • AX265 PathCamHD Camera
  • AX266 PathCamHR Camera
  • AX102 PathCam Micron Microscope
Specifications
Product Type Grossing Station
For Use With (Equipment) Foot Pedal