Learn More
Mopec PathCam Micron Maestro Mount
SDP
Catalog No. 21377355 Shop All Mopec Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
21-377-355 Stainless Steel
1 options
Catalog No. 21-377-355 Supplier Mopec Supplier No. AX133
Add to Cart
Add to Cart
Designed to attach your PathCam Micron camera to your Maestro Encore grossing station.
  • Precision machine crafted components provide smooth adjustment and positioning
  • Easy to use-simple installation and effortless operation
  • Integrated cable management
  • Balanced and stable ergonomic design
  • Maestro and Maestro Encore grossing stations
  • AX250 PathCam Camera Wall-Mount
Specifications
Product Type Grossing Station
Material Stainless Steel