Learn More
Mopec PathCam Micron Microscope
SDP
Catalog No. 23987019 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Micron Camera
Catalog No. For Use With (Equipment)
23-987-019 Micron Camera
1 options
Catalog No. 23-987-019 Supplier Mopec Supplier No. AX102
Add to Cart
Add to Cart
Delivers unmatched magnification and resolution, allowing ultra close visualization of minute structure.
  • Preprogrammed with PathCam Software settings: 1.3 MP CMOS Image Sensor
  • Magnified high-resolution capture
  • Annotate and label minute structures in software
  • Manual focus
  • Auto White Balance
  • Auto Exposure
  • (8) Built-in LED white lights
  • Adjustable polarizer: reveals details by reducing reflections
  • Measures distance and area in microns
  • Add customizable grid in image area
  • Fits all Mopec Maestro and Maestro Encore grossing stations
  • Can also be used on AX260 Mobile Imaging Station (pictured)
Specifications
Product Type Grossing Station
For Use With (Equipment) Micron Camera