Learn More
Mopec PathCam Monitor
SDP
Catalog No. 23987016 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Monitor
Catalog No. For Use With (Equipment)
23-987-016 Monitor
1 options
Catalog No. 23-987-016 Supplier Mopec Supplier No. AX045
Add to Cart
Add to Cart
Ideal solution to add to your Maestro Encore grossing station to view your PathCam Imaging Software.
  • LED full HD display
  • 1920 x 1080 pixels
  • 10-point multi-touch interactivity
  • Inputs: DisplayPort, VGA, and HDMI
  • (2) integrated speakers
  • VESA mount compatible
  • Blue light reduction
  • HID compliant
  • Maestro and Maestro Encore grossing stations equipped with PathCam Imaging Systems
Specifications
Product Type Grossing Station
For Use With (Equipment) Monitor