Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymedco PATHFAST Immunoassay Reagent Myoglobin II
SDP

Catalog No. 21200133 Shop All Polymedco Products
Change view
Click to view available options
No. of Reactions:
60
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
21-200-133 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-133 Supplier Polymedco Supplier No. PF1111K
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Myoglobin reagent and calibrators

  • Includes 60 tests

Specifications

CE Marker CE-IVD
Clia Complexity Moderate (Serum, Plasma or Whole Blood), Waived (Other)
Disposable Single-use
No. of Reactions 60
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.