Learn More
Eurofins Discoverx PathHunter Flash Detection Kit, 25 (2 plates), 200/800 dp (96-well/384-well). (93-0247M)
Catalog No. NC1310618
Encompass
The PathHunter Flash Detection Kit is a detection kit intended for use with PathHunter cell lines or with custom cell lines made using ProLabel/PK expression vectors. This kit will provide a brighter signal with shorter kinetics than the PathHunter Detection Kit, at the cost of a shorter half life. It can be substituted in and used with any cell line that specifies use of the PathHunter Detection kit. This kit contains lysis buffer and EFC substrate to allow for detection of EFC levels in the assayed cells. This kit contains all components required to assay 2 plates (96/384-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0247m.
Catalog No. NC1310618 Supplier Eurofins Discoverx Supplier No. 930247M
The PathHunter Flash Detection Kit is a detection kit intended for use with PathHunter cell lines or with custom cell lines made using ProLabel/PK expression vectors. This kit will provide a brighter signal with shorter kinetics than the PathHunter Detection Kit, at the cost of a shorter half life. It can be substituted in and used with any cell line that specifies use of the PathHunter Detection kit. This kit contains lysis buffer and EFC substrate to allow for detection of EFC levels in the assayed cells. This kit contains all components required to assay 2 plates (96/384-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0247m.