Learn More
StatLab PathFlow Connected Lid Cassettes Biopsy
SDP
Catalog No. 21402134
Change view
Click to view available options
Pore Style:
Loose
Taped
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Style
21-402-134 Loose
21-402-135 Taped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-134 Supplier StatLab Supplier No. CAS506PC
Add to Cart
Add to Cart
Cassettes biopsy , one piece design with rear hinge and front tab closure
  • The 35 degree print face is designed to be compatible for printing on the Leica IP C Inkjet Cassette Printer, Primera Signature cassette printer and the Tissue-Tek SmartWrite Cassette Printer
  • These cassettes are a suitable replacement for the Unisette (M506) and Signature cassettes(M406)
  • Loose,1500 per case
Specifications
Pore Style Loose
Color Peach
Product Type Biopsy Cassette