Learn More
StatLab PathFlow Connected Lid Cassettes Biopsy
Description
- The 35 degree print face is designed to be compatible for printing on the Leica IP C Inkjet Cassette Printer, Primera Signature cassette printer and the Tissue-Tek SmartWrite Cassette Printer
- These cassettes are a suitable replacement for the Unisette (M505) and Signature cassettes(M405)
- Loose, 1500/case
- Taped, 1000/case
Specifications
Specifications
| Pore Style | Biopsy Loose |
| Color | Aqua |
| Product Type | Biopsy Cassettes |