Learn More
StatLab PathFlow Connected Lid Cassettes Biopsy
SDP
Catalog No. 21033316
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Tan
White
Yellow
Pore Style:
Biopsy Loose
Biopsy Taped
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Pore Style
21-033-316 Yellow Biopsy Taped
21-033-297 Aqua Biopsy Loose
21-033-298 Aqua Biopsy Taped
21-033-299 Blue Biopsy Loose
21-033-300 Blue Biopsy Taped
21-033-301 Green Biopsy Loose
21-033-302 Green Biopsy Taped
21-033-303 Gray Biopsy Loose
21-033-304 Gray Biopsy Taped
21-033-305 Lilac Biopsy Loose
21-033-306 Lilac Biopsy Taped
21-033-307 Orange Biopsy Loose
21-033-308 Orange Biopsy Taped
21-033-309 Pink Biopsy Loose
21-033-310 Pink Biopsy Taped
21-033-311 Tan Biopsy Loose
21-033-312 Tan Biopsy Taped
21-033-313 White Biopsy Loose
21-033-314 White Biopsy Taped
21-033-315 Yellow Biopsy Loose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 21-033-316 Supplier StatLab Supplier No. CAS506YLT
Add to Cart
Add to Cart
One Piece design with rear hinge and front tab closure with loose, 1500 per case
  • The 35 degree print face is designed to be compatible for printing on the Leica IP C Inkjet Cassette Printer, Primera Signature cassette printer and the Tissue-Tek SmartWrite Cassette Printer
  • These cassettes are a suitable replacement for the Unisette (M505) and Signature cassettes(M405)
  • Loose, 1500/case
  • Taped, 1000/case
Specifications
Pore Style Biopsy Taped
Color Yellow
Product Type Biopsy Cassettes