Learn More
StatLab PathFlow PiSmart Laser Attached Lid Front Hinge Biopsy Cassettes
SDP
Catalog No. 21200149
Change view
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-200-149 Yellow
21-200-141 Blue
21-200-142 Green
21-200-143 Gray
21-200-144 Lavender
21-200-145 Orange
21-200-146 Pink
21-200-147 Tan
21-200-148 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 21-200-149 Supplier StatLab Supplier No. CASLPI200BXYL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Threaded biopsy cassettes with attached lid and front hinge.
  • 0.9 mm pores
  • Front hinge with lid attached
  • Made from acetal polymer
  • Specifically validated for the Pi Smart Laser Cassette Printer
  • Print face features four raised pips in the corners to protect printed surface during dewaxing, while enabling the surface to be cleared of wax
  • Threaded
  • 2000 per case
Specifications
Quantity 2000 Cassettes/Cs.
Color Yellow
Product Type Biopsy Cassettes
Made in USA