Learn More
StatLab PathFlow PiSmart Slotted Thermal Transfer Embedding Cassettes
SDP
Catalog No. 21402271
Change view
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-402-271 Blue
21-402-272 Green
21-402-273 Gray
21-402-274 Lavender
21-402-275 Orange
21-402-276 Pink
21-402-277 Tan
21-402-278 White
21-402-279 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 21-402-271 Supplier StatLab Supplier No. CASPI100SLBL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
PathFlow PiSmart Slotted Thermal Transfer Embedding Cassettes, Threaded with Lid.
  • 1,000/bx, 2 bx/case, 2,000/cs
  • Threaded in strips of 40 with lids separate
  • PathFlow PiSmart Cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing
  • Engineered specifically for use in the PiSmart Thermal Transfer Cassette printer
  • The new and improved cassettes now incorporate a smoother print face, resulting in improved print of barcodes and text
  • Available in 9 colors
Specifications
Color Blue
Product Type Embedding Cassettes
Made in USA