Learn More
StatLab PathFlow PiSmart Thermal Transfer Attached Lid Front Hinge Microbiopsy Cassettes
SDP
Catalog No. 21200177
Change view
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-200-177 Blue
21-200-178 Green
21-200-179 Gray
21-200-180 Lavender
21-200-181 Orange
21-200-182 Pink
21-200-183 Tan
21-200-184 White
21-200-185 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 21-200-177 missing translation for 'mfr' StatLab missing translation for 'supplierNo' CASPI200MBXBL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermal transfer microbiopsy cassettes with attached lid and front hinge.
  • 0.38 mm pores
  • Front hinge with lid attached
  • Made from acetal polymer
  • Specifically validated for the Pi Smart Thermal Transfer Cassette Printer
  • Print face features four raised pips in the corners to protect printed surface during dewaxing, while enabling the surface to be cleared of wax
  • Threaded, 2000 per case
Specifications
Quantity 2000 Cassettes/Cs.
Color Blue
Product Type Microbiopsy Cassettes
Made in USA