Learn More
StatLab PathFlow PiSmart Thermal Transfer Attached Lid Front Hinge Microbiopsy Cassettes
SDP
Catalog No. 21200185
Change view
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-200-185 Yellow
21-200-177 Blue
21-200-178 Green
21-200-179 Gray
21-200-180 Lavender
21-200-181 Orange
21-200-182 Pink
21-200-183 Tan
21-200-184 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 21-200-185 Supplier StatLab Supplier No. CASPI200MBXYL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermal transfer microbiopsy cassettes with attached lid and front hinge.
  • 0.38 mm pores
  • Front hinge with lid attached
  • Made from acetal polymer
  • Specifically validated for the Pi Smart Thermal Transfer Cassette Printer
  • Print face features four raised pips in the corners to protect printed surface during dewaxing, while enabling the surface to be cleared of wax
  • Threaded, 2000 per case
Specifications
Quantity 2000 Cassettes/Cs.
Color Yellow
Product Type Microbiopsy Cassettes
Made in USA