Learn More
StatLab PathFlow PiSmart Thermal Transfer Attached Lid Front Hinge Slotted Embedding Cassettes
SDP
Catalog No. 21200194
Change view
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-200-194 Yellow
21-200-186 Blue
21-200-187 Green
21-200-188 Gray
21-200-189 Lavender
21-200-190 Orange
21-200-191 Pink
21-200-192 Tan
21-200-193 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 21-200-194 Supplier StatLab Supplier No. CASPI200SLYL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Slotted embedding thermal transfer cassettes with attached lid and front hinge.
  • Slotted embedding cassettes
  • Front hinge with lid attached
  • Made from acetal polymer
  • Specifically validated for the Pi Smart Thermal Transfer Cassette Printer
  • Print face features four raised pips in the corners to protect printed surface during dewaxing, while enabling the surface to be cleared of wax
  • Threaded
  • 2000 per case
Specifications
Quantity 2000 Cassettes/Cs.
Color Yellow
Product Type Slotted Embedding Cassettes
Made in USA