Learn More
StatLab PathFlow Supa Mega Slide Rack Adapter
SDP
Catalog No. 22000075
Change view
Click to view available options
Product Type:
Staining Rack System
Catalog No. Product Type
22-000-075 Staining Rack System
1 options
Catalog No. 22-000-075 Supplier StatLab Supplier No. RMC200063A
Add to Cart
Add to Cart
The Slide Rack Adapter enables the user to stain Supa Mega slides
  • The Slide Rack Adapter enables the user to stain Supa Mega slides on automated staining equipment bringing convenience, efficiency and cost savings all at the same time
  • Stainless Steel
Specifications
Product Type Staining Rack System
For Use With (Equipment) Automated Staining Equipment