Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab PathFlow SupaMega Base Molds
SDP

Catalog No. 21033376
Change view
Click to view available options
Length (Metric):
36 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
21-033-376 36 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-376 Supplier StatLab Supplier No. GBC361405A

missing translation for 'validMainframeMsg'

Stainless steel designed for a lifetime of use with re-usable feature

  • The Supa Mega Stainless Steel Base Mold has extra deep side walls which allow wax to be decanted into the cassette without the risk of the usual leakage of wax
  • Re-usable
  • Stackable reducing storage space

Specifications

Product Type Base Mold
Material Stainless Steel
Color Gray
Length (Metric) 36 m
Width (Metric) 36 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.