Learn More
StatLab PathFlow SupaMega MotherShip Cassettes
SDP
Catalog No. 21033365
Change view
Click to view available options
Color:
Red
Catalog No. Color
21-033-365 Red
1 options
Catalog No. 21-033-365 Supplier StatLab Supplier No. EAO011002A
Add to Cart
Add to Cart
An innovative solution for processing large tissue specimens
  • The Supa Mega Mothership cassette uses an innovative permanent docking bay for the majority of standard-sized tissue processing cassettes printed with ID bar codes and/or test information
  • This enables the specimen to be entered onto a LIMS guaranteeing its accurate and reliable tracking
  • Enables specimens to be entered onto Laboratory Information System (LIS)
  • Better reagent flow and improved turnaround times
  • Casting done in 30 minutes at -8°C
  • Mothership Cassette can accommodate the majority of available standard-sized cassettes
  • Large color range available with bespoke colors available on request
  • 100 per box
Specifications
Color Red
Product Type Large Embedding Cassette