Learn More
StatLab PathFlow SupaMega Slim Cassettes
SDP
Catalog No. 22222130
Change view
Click to view available options
Color:
Blue
Pink
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-222-130 White
21-033-361 Blue
21-033-362 Pink
21-033-363 Yellow
21-033-364 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22-222-130 Supplier StatLab Supplier No. EAN010202A
 Restricted Item
Add to Cart
Add to Cart
Thin mega cassettes allow for speedy processing of large format tissue specimens
  • SupaMega Slim cassettes uses 1/3 the depth of standard SupaMega Cassettes
  • Better reagent flow and improved turnaround times
  • Casting can be done in 20 minutes at -8°C
  • Less wax used during the embedding process
  • Large color range available
  • Specific base molds
  • 100 per box
  • Base dimensions: 52.3 mm x 75 mm x 9.5 mm
Specifications
Color White
Product Type Large Embedding Cassette