Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab PathFlow Universal Supa Mega Cassette Clamp
SDP

Catalog No. 22000045
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-000-045 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-000-045 Supplier StatLab Supplier No. JFB010000A
Add to Cart
Add to Cart

The Supa Mega Cassette Clamp is used for the microtomy of large tissue blocks

  • Incorporating an intuitive quick-release system, the clamp can securely hold any tissue processing cassette from our Supa Mega range
  • Unlike other large tissue block clamps, the Supa Mega Clamp's unique design allows sectioning all the way through the Supa Mega block without collision between the clamp and knife block
  • The design also prevents the possibility of the cassette being attached in the wrong orientation
  • Easily fits Leica Biosystems' HistoCore, RM2100 and RM2200 and Epredia's Microm HH series of microtomes

Specifications

Product Type Cassette Clamp
For Use With (Equipment) Epredia Microtomes
Disposable Reusable
Color Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.