Learn More
Eurofins Discoverx PathHunter Detection Kit, 550 (100 plates), 40,000 dp (384-well). (93-0001L)
Catalog No. NC9661103
Encompass
The PathHunter Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter cell lines or with custom cell lines made using ProLabel/PK expression vectors. This kit contains lysis buffer and EFC substrate to allow for detection of EFC levels in the assayed cells. This kit contains all components required to assay 100 plates (96/384 well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0001l.
Catalog No. NC9661103 Supplier Eurofins Discoverx Supplier No. 930001L
Add to Cart
Add to Cart
Request a Quote
The PathHunter Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter cell lines or with custom cell lines made using ProLabel/PK expression vectors. This kit contains lysis buffer and EFC substrate to allow for detection of EFC levels in the assayed cells. This kit contains all components required to assay 100 plates (96/384 well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0001l.