Learn More
Eurofins Discoverx PathHunter eXpress CSF1R/CSF1R Dimerization Assay, 2pl Kit. (93-1061E3CP5M)
Catalog No. NC1639735
Encompass
The PathHunter CSF1R/CSF1R Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. M-CSF) induced dimerization of CSF1R/CSF1R receptors. This product uses EFC technology. The cell line included overexpresses CSF1R target protein fused to PK alongside CSF1R target protein fused to EA. Upon ligand-induced dimerization of CSF1R/CSF1R, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. NC1639735 Supplier Eurofins Discoverx Supplier No. 931061E3CP5M
The PathHunter CSF1R/CSF1R Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. M-CSF) induced dimerization of CSF1R/CSF1R receptors. This product uses EFC technology. The cell line included overexpresses CSF1R target protein fused to PK alongside CSF1R target protein fused to EA. Upon ligand-induced dimerization of CSF1R/CSF1R, detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.