Learn More
Eurofins Discoverx PathHunter eXpress GLP1R Total GPCR Internalization Assay, 2-Plate Kit. (93-1075E3CP5M)
Catalog No. NC1603545
Encompass
The PathHunter GLP1R Total GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit uses EFC technology to measure ligand-induced internalization of GLP1R via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged GLP1R and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the GLP1R receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.
Catalog No. NC1603545 Supplier Eurofins Discoverx Supplier No. 931075E3CP5M
The PathHunter GLP1R Total GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit uses EFC technology to measure ligand-induced internalization of GLP1R via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged GLP1R and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the GLP1R receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. For detailed product information, User Manual and Datasheet, visit www.discoverx.com/express.