Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab PathMark Margin Marker
SDP

Catalog No. 21033444
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-033-444 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-444 Supplier StatLab Supplier No. MA0116005

missing translation for 'validMainframeMsg'

Tissue marking dye with 2 oz. bottle, avaialble in orange color

  • 2 oz. bottle
  • Orange
  • Tissue Marking Dye

Specifications

Color Orange
Product Type Tissue Marking Dye
Solution Type Mordant Dye
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.