Learn More
Mopec PathOdorizer Odor Eliminating Solution
SDP
Catalog No. p-7116290 Shop All Mopec Products
Change view
Click to view available options
Description:
Deodorizer
Deodorizer, Cleaner and Degreaser
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
22-444-302 Deodorizer
22-444-303 Deodorizer, Cleaner and Degreaser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-444-302 Supplier Mopec Supplier No. BE040
Only null left
Add to Cart
Add to Cart
Mopec PathOdorizer Odor Eliminating Solution removes odors from clothes, vehicles, body bags, refrigerators and other items
  • Eliminates offensive odors emitted from decomposed bodies within seconds
  • Developed specially for use by medical examiners
  • Can be used in every phase of work from the discovery scene to the morgue
  • Removes odors from clothes, vehicles, body bags, refrigerators and other items
  • Keeps the odor from permeating the lab environment
  • Water soluble
  • Odorless
  • Colorless
  • Biodegradable
  • Non-flammable
  • Non-corrosive
Specifications
Appearance Colorless
Description Deodorizer