Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PathoDx Strep Grouping Universal Kit
GSA_VA

Catalog No. R62076 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
60 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R62076 60 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R62076 Supplier Thermo Scientific™ Supplier No. R62076
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Support the functionality of the Thermo Scientific™ PathoDx™ Strep Grouping Kit with Thermo Scientific™ PathoDx™ Strep Grouping Universal Kit.

Support the functionality of the Thermo Scientific™ PathoDx™ Strep Grouping Kit with Thermo Scientific™ PathoDx™ Strep Grouping Universal Kit. This kit is a set of components to be used with selected, separately purchased PathoDxStrep Grouping Reagents designed to identify beta-haemolytic streptococci of Lancefield Groups A, B, C, F or G from primary culture plates.This accessory kit contains: Extraction reagent 1; Extraction reagent 2; Extraction reagent 3; Positive control; Reaction cards; Stirrers.

Includes:

Control antigens ABC and FG; extraction reagents 1, 2, 3; stirrers; disposable slides

Requires:

For use with Part No. R62030, R62031, R62032, R62034, R62035.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive
Description Strep Universal Grouping Kit
Detectable Analytes beta-haemolytic Streptococci (Groups A, B, C, F, G)
Type Streptococcus Test Kit
Includes Control antigens ABC and FG; extraction reagents 1, 2, 3; stirrers; disposable slides
Organism Group Lancefield A, B, C, D, F and G
Quantity 60 Tests/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.