Learn More
Zymo Research Corporation Pathogen DNA/RNA Buffer ( 50 ml)
SDP
Catalog No. 502037244
Encompass_Preferred
Pathogen DNA/RNA Buffer is an all-in-one lysis and binding reagent used in combination with DNA/RNA Shield to facilitate the isolation of pathogen (virus, bacteria, protozoa) DNA and RNA from a wide variety of vectors (mosquitoes, fleas, ticks, etc.) and tissue types (mammals, birds, etc.).
Catalog No. 50-203-7244 Supplier Zymo Research Corporation Supplier No. R1042150
Pathogen DNA/RNA Buffer is an all-in-one lysis and binding reagent used in combination with DNA/RNA Shield to facilitate the isolation of pathogen (virus, bacteria, protozoa) DNA and RNA from a wide variety of vectors (mosquitoes, fleas, ticks, etc.) and tissue types (mammals, birds, etc.).