Learn More
Zymo Research Corporation Pathogen DNA/RNA Wash Buffer (Concentrate), 48 ml
SDP
Catalog No. 502037245
Encompass_Preferred
Pathogen DNA/RNA Wash Buffer is an all-in-one lysis and binding reagent used in combination with DNA/RNA Shield to facilitate the isolation of pathogen (virus, bacteria, protozoa) DNA and RNA from a wide variety of vectors (mosquitoes, fleas, ticks, etc.) and tissue types (mammals, birds, etc.).
Catalog No. 50-203-7245 Supplier Zymo Research Corporation Supplier No. R1042248
Pathogen DNA/RNA Wash Buffer is an all-in-one lysis and binding reagent used in combination with DNA/RNA Shield to facilitate the isolation of pathogen (virus, bacteria, protozoa) DNA and RNA from a wide variety of vectors (mosquitoes, fleas, ticks, etc.) and tissue types (mammals, birds, etc.).