Learn More
Microscope World Fein Optici RB40 Pathology Lab Microscope
SDP
Supplier:  Microscope World RB41PATHERGOTRINOC
Encompass
Quintuple objective turret holds the included DIN Infinity Corrected Optical System (ICOS) ObjectivesPlan Achromat 2x, NA 0.06, WD 5mmPlan Semi-Apochromat Fluor 4x, NA 0.13, wD 18.5mmPlan Semi-Apochromat Fluor 10x, NA 0.30, WD 10.6mmPlan Semi-Apochromat Fluor 20x, NA 0.50, WD 2.33mmPlan Semi-Apochromat Fluor 40x, NA 0.75, WD 0.6mmErgonomic trinocular head with erect image adjusting 5-35deg with beam splitter positions of 100-0, and 0-100, FN25.FPL-WF 10x Widefield, high eyepoint eyepieces, FN22.Magnification- 20x, 40x, 100x, 200x and 400x.Double-layer mechanical stage, 175mm x 145mm with right-hand ergonomic controls. Specimen holder allows for either one or two slides. X-Y Axis travel of 76mm x 50mm.5W LED transmitted Koehler light system. Telecentric Koehler illumination system provides NA 1.2/0.22 swing-out condenser for use with 2x-100x objectives. Coaxial coarse and fine focus adjustment with graduations of 2um. 250mm W x 463mm D x 408mm HWarranty- 5 Years.
Catalog No. NC2965681
Add to Cart
Request a Quote